Twaddell Hydrometers
Twaddell Hydrometers
Actual product may vary
Fisherbrand
Twaddell Hydrometers
For reporting the measured specific gravity of a liquid relative to water. Brannan™ Twaddell Hydrometers are manufactured from soda glass, steel shot ballast and synthetic wax resin.
Have Questions?
Catalog NumberSpecific Gravity Range
FKS1285917774 to 102 Tw
Catalog number FKS12859177
Price (SGD)
53.00
EA
Add to cart
Specific Gravity Range:
74 to 102 Tw
Price (SGD)
53.00
EA
Add to cart
For reporting the measured specific gravity of a liquid relative to water. Brannan™ Twaddell Hydrometers are manufactured from soda glass, steel shot ballast and synthetic wax resin.
  • 160 mm in length
  • With single degree divisions
  • Adjusted for use at 15.6°C (60°F)
  • For medium surface tension liquids
  • Available for numerous Twaddell ranges
Specifications
Quantity1 Ea.
TypeTwaddle Hydrometer
Accuracy±1 Tw
Increments1 Tw
Length (Metric)160 mm
Ballast TypeSteel Shot
Specific Gravity Range74 to 102 Tw
Unit SizeEA