Check Order Status
Cork Borers
Actual product may vary
Fisherbrand
Cork Borers
Efficiently and cleanly bores holes in cork. Chroma Gesellschaft™ Cork Borers have a cutting tube mounted with safety under-flange in shaped handle.
Have Questions?
Catalog Number
Material
FKS12853952
Brass
Catalog number
FKS12853952
Price (SGD)
251.00
EA
Add to cart
Material:
Brass
Price (SGD)
251.00
EA
Add to cart
With rod for clearing bores
Manufactured from brass
Sizes numbered consecutively from 1 upwards in each set
Specifications
Product Type
Cork Borer
Material
Brass
For Use With (Application)
For boring holes in cork.
Diameter (Metric)
4 to 10 mm