Standard Series™ 10mm Precision NMR Sampling Tubes
Standard Series™ 10mm Precision NMR Sampling Tubes
Actual product may vary
Fisherbrand
Standard Series™ 10mm Precision NMR Sampling Tubes
Excellent for monitoring reactions, scouting for new compounds or doing routine NMR. Norell™ Standard Series™ 10mm Precision NMR Sampling Tubes have been developed for quality and precision and are the top choice for today's scientist.
Have Questions?
Catalog NumberLength (Metric)Diameter (Metric) Outer
FKS12852296178 mm4.97 mm
Catalog number FKS12852296
Price (SGD)
347.00
PK
Add to cart
Length (Metric):
178 mm
Diameter (Metric) Outer:
4.97 mm
Price (SGD)
347.00
PK
Add to cart
  • For auto-sampling systems and high-throughput NMR
  • Polished ends to make tubes less susceptible to breaking
  • Colored caps included
  • Virtually all bottom and wall variation has been eliminated, greatly improving shimmering times and sample reproducibility
Specifications
Length (Metric)178 mm
MaterialBorosilicate Glass
Product TypeUltra Precision NMR Sample Tube
Diameter (Metric) Outer4.97 mm
Diameter (Metric) Inner4.2 mm
For Use With (Application)Routine NMR
IncludesCap
Quantity5 Pack
Unit SizePK