Check Order Status
Cylinder Brush
Actual product may vary
Fisher Catalogue
Cylinder Brush
Manufactured with galvanized steel handle. Pinnacle Brush™ Cylinder Brush features nylon bristles.
Have Questions?
Catalog Number
Length (Metric)
FKS12851944
685 mm
Catalog number
FKS12851944
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric):
685 mm
Fan-shaped end
Galvanised steel wire handle
Specifications
Product Type
Cylinder Brush
Bristle Material
Nylon
Handle Material
Steel Wire
Diameter (Metric) Bristle
76 mm
Length (Metric)
685 mm
Length (Metric) Brush
170 mm
Quantity
10 Pk.
Size
76 x 170 mm