Microscope Slides with Cavities
Microscope Slides with Cavities
Actual product may vary
Fisherbrand
Microscope Slides with Cavities
Use Marienfeld Superior™ Soda Lime Glass Microscope Slides with carefully ground and polished cavities for research needs in the laboratory.
Have Questions?
Catalog NumberWell Count
FKS128487152
Catalog number FKS12848715
Price (SGD)
485.00
PK
Add to cart
Well Count:
2
Price (SGD)
485.00
PK
Add to cart
  • Premier quality microscope slides with cavities
  • Made of soda lime glass of third hydrolytic class
  • With 90° ground edges
  • Cavities with diameter of 15-18 mm and a depth of 0.6-0.8 mm
  • Pre-cleaned, ready for use, autoclavable
  • For in-vitro diagnostic (IVD) applications according to IVD directive 98/79/EC, with CE-mark, recommended best-before-date and batch number for comprehensive information and traceability
Specifications
No. per Pack50
Edges90°, Ground
FrostedNot Frosted
Length (Metric)76 mm
MaterialSoda Lime Glass Type III, Hydrolytic Class
PrecleanedYes
Thickness (Metric)1.2 to 1.5 mm
TypeMicroscope Slide
Well Count2
Width (Metric)26 mm
DescriptionCavity Slides
Quantity50 Pack
Dimensions (L x W)76 x 26 mm
Unit SizePK