Search Thermo Fisher Scientific
Catalog Number | Diameter (Metric) Inside | Gauge |
---|---|---|
FKS12840155 | 5.05 mm | Heavy |
FKS11919054 | 1.59 mm | Standard |
An inoculation loop is a simple tool used to gather and transfer microscopic organisms from a culture or broth plate. The small loop removes a consistent amount of inoculum by using surface tension. The inoculation loop is flame-sterilized before and after each use to eliminate cross-contamination between cultures. (The loop must be cooled after sterilization to avoid destroying the organisms.)