Polymix™ PX-MFC 90 D Hammer or Cutting Mill
Polymix™ PX-MFC 90 D Hammer or Cutting Mill
Actual product may vary
Fisherbrand
Polymix™ PX-MFC 90 D Hammer or Cutting Mill
Polymix™ MFC 90 D features easy to change grinding attachments and sieves extending the range of samples that can be processed. Simple handling, high user safety and efficient grinding results are just a few of the advantages of this mill.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypePlug Type
FKS12839035Blade Grinding MillUK plug
FKS12829035Hammer Grinding MillGB plug
Catalog number FKS12839035
Price (SGD)
41,702.00
EA
Add to cart
Product Type:
Blade Grinding Mill
Plug Type:
UK plug
Price (SGD)
41,702.00
EA
Add to cart
Polymix™ MFC 90 D features easy to change grinding attachments and sieves extending the range of samples that can be processed. Simple handling, high user safety and efficient grinding results are just a few of the advantages of this mill.
  • Working volume approx. 300mL (volume of the funnel)
  • 600W high frequency 3 phase motor with speed regulation from 50rpm up to 6, 000rpm
  • Integrated overheating, overload and blocking protection
  • Safety switch for automatic off
  • Energy saving through automatic standby mode
  • Hammer grinding or blade grinding attachment
  • Sieves in different mesh sizes from 0.2mm up to 6mm Suitable for all dry milling applications in the laboratory, including dry milling of dried, non-fatty, stringy and ductile products such as wood, bark, roots, leaves, straw, cork etc. and dried, non-fatty, hard and brittle materials such as cereals, grain, malt, raw and roasted coffee, beans, oats, nutshells, amber, mortar, etc.
Specifications
Product TypeBlade Grinding Mill
Certifications/ComplianceIEC/EN 61000-6-2 / EN 61000-6-3, IEC/EN 61010-2-51
Dimensions (L x W x H)325 x 251 x 480 mm
Electrical Requirements230 V/GB, 50 Hz, 1'000 W Motor Input Power
Height (Metric)480 mm
Hertz50/60 Hz
IP RatingIP20
Length (Metric)325 mm
Plug TypeUK plug
Voltage230 V
Watts1000 W
Weight (Metric)12 kg
Width (Metric)251 mm
Unit SizeEA