BMP™ Label Cartridges for BMP21-PLUS; BMP21-LAB; BMP21; LABPAL, clear polyester film tape
BMP™ Label Cartridges for BMP21-PLUS; BMP21-LAB; BMP21; LABPAL, clear polyester film tape
Actual product may vary
BMP™ Label Cartridges for BMP21-PLUS; BMP21-LAB; BMP21; LABPAL, clear polyester film tape
BMP™ Label Cartridges for BMP21-PLUS; BMP21-LAB; BMP21; LABPAL, clear polyester film tape
BMP™ Label Cartridges for BMP21-PLUS; BMP21-LAB; BMP21; LABPAL, clear polyester film tape
Fisherbrand
BMP™ Label Cartridges for BMP21-PLUS; BMP21-LAB; BMP21; LABPAL, clear polyester film tape
Use this film tape for a variety of labeling applications. BMP™ Label Cartridges for BMP21-PLUS; BMP21-LAB; BMP21; LABPAL are ideal for centrifuge, Eppendorf and PCR tubes; conical and large tubes; cryo tubes and vials.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberWidth (English)
FKS128372680.3 in.
FIS19-102-7180.3 in.
FIS19-102-7310.5 in.
FIS19-102-7440.75 in.
Catalog number FKS12837268
Price (SGD)
160.00
EA
Add to cart
Width (English):
0.3 in.
Price (SGD)
160.00
EA
Add to cart
Use this film tape for a variety of labeling applications. BMP™ Label Cartridges for BMP21-PLUS; BMP21-LAB; BMP21; LABPAL are ideal for centrifuge, Eppendorf and PCR tubes; conical and large tubes; cryo tubes and vials.
  • High adhesive
  • Translucent
  • Gloss finish
  • Perfect for autoclaving and for use in below freezing temperatures
Specifications
Certifications/ComplianceHalogen-free (DIN VDE 0472/parts 815)
Adhesive TypePermanent Acrylic
ColorBlack, White
FinishMatte
FormatCartridge (Label)
MaterialPolyester
Product TypeLabel Cartridge
For Use With (Application)General Identification, Laboratory Identification: Vial, Well Plate
For Use With (Equipment)M210 Label Printer, M210-LAB Label Printer, M211 Label Printer, BMP™21 LAB Label Printer, BMP™21-PLUS Label Printer
Quantity1 Roll
Length (English)21 ft.
Width (English)0.3 in.
Height (English)21 ft.
Height (Metric)6.4 cm
Alternate Part NumberM21-375-461-AW
Unit SizeEA