Chromabond™ PTL Separation Column
Chromabond™ PTL Separation Column
Actual product may vary
Fisherbrand
Chromabond™ PTL Separation Column
Separate compounds with gravity flow phase separation. Chromabond™ PTL Separation Columns come in a variety of capacities for solvents lighter than water.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS1283430115 mL
FKS128110316 mL
Catalog number FKS12834301
Price (SGD)
1,310.00
PK
Add to cart
Capacity (Metric):
15 mL
Price (SGD)
1,310.00
PK
Add to cart
  • Automatic separation of a two-phase mixture without separation funnel
  • Two-phase mixtures are completely applied to the column and the phase boundary is determined without further work.
  • The special membrane automatically stops the flow when the lower phase has passed.
  • The upper phase remains in the column, thus both phases are available for futher analysis.
  • Columns must not be run with vacuum or pressure
  • PTL for solvents lighter than water, e.g., for diethyl ether, hexane maximum size 70 mL
Specifications
Capacity (Metric)15 mL
MaterialPTL
Quantity100 Pk.
For Use With (Application)Gravity Flow Phase Separation
TypePhase Separation Column
Sample Volume15 mL
Unit SizePK