13mm LPG Bunsen Burner
13mm LPG Bunsen Burner
Actual product may vary
Fisherbrand
13mm LPG Bunsen Burner
An excellent solution for heating, sterilization and combustion in the laboratory. R&L Enterprises™ 13mm LPG Bunsen Burners have a typical gas consumption of 107.5L per hour.
Have Questions?
Catalog NumberHeight (Metric)
FKS12832164145 mm
Catalog number FKS12832164
Price (SGD)
386.00
PK
Add to cart
Height (Metric):
145 mm
Price (SGD)
386.00
PK
Add to cart
  • For use with propane and butane gases
  • Temperature is achieved at the tip of the blue flame cone
  • Constructed using a heavy pressure diecast base painted in chemically resistant blue acrylic finish
  • Tested for flame quality and leakage prior to packing
Specifications
Product TypeBunsen Burner
Gas TypeButane, Propane, LPG
Diameter (Metric) Outer Inlet13 mm
Height (Metric)145 mm
No. per Pack5 Pk.
Weight (Metric)0.8 kg
Unit SizePK