Check Order Status
Platinum Crucible
Actual product may vary
Fisherbrand
Platinum Crucible
Available in 15mL and 50mL capacities
Have Questions?
Catalog Number
Capacity (Metric)
FKS12827673
50 mL
Catalog number
FKS12827673
Price (SGD)
12,308.00
EA
Add to cart
Capacity (Metric):
50 mL
Price (SGD)
12,308.00
EA
Add to cart
Quoted masses are approximate.
Approximate process temperature 1,200°C and maximum temperature is 1,400°C (for short time of 10min)
Specifications
Material
Platinum
Max. Temperature (Metric)
1200°C
Quantity
1Each
Capacity (Metric)
50 mL
Diameter (Metric) Outer
45mm
Height (Metric)
50mm
Product Type
Crucible