Stainless Steel Jugs
Stainless Steel Jugs
Actual product may vary
Fisher Catalogue
Stainless Steel Jugs
304 spec stainless steel jugs are suitable for pharmaceutical and laboratory applications.
Have Questions?
Catalog NumberCapacity (Metric)
FKS12827595500 mL
Catalog number FKS12827595
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
500 mL
  • 304 spec stainless steel seamless jugs
  • Suitable for pharmaceutical and laboratory applications
Specifications
Diameter (Metric) Outer90 mm
Capacity (Metric)500 mL
MaterialStainless Steel
FinishBright
Height (Metric)135 mm
TypeJug