1x1 image pixel for data collection
BS 718 M100 Density Hydrometers
BS 718 M100 Density Hydrometers
Actual product may vary
Fisherbrand
BS 718 M100 Density Hydrometers
For reporting the measured specific gravity of a liquid relative to water. Brannan™ BS 718 M100 Density Hydrometers are manufactured from soda glass, steel shot ballast and synthetic wax resin.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSpecific Gravity RangeIncludes
FKS128192071.200 to 1.300 g/mLPlastic protective tube
FKS128092071.100 to 1.200 g/mLPlastic protective tube
Catalog number FKS12819207
Price (SGD)
64.00
EA
Estimated availability date 01-Oct-2025
Add to cart
Specific Gravity Range:
1.200 to 1.300 g/mL
Includes:
Plastic protective tube
Price (SGD)
64.00
EA
Add to cart
For reporting the measured specific gravity of a liquid relative to water. Brannan™ BS 718 M100 Density Hydrometers are manufactured from soda glass, steel shot ballast and synthetic wax resin.
  • BS 718 : 1991 is generally aligned with ISO 387 and ISO 649
  • With a range span of 0.100g/mL
  • Available for numerous specific gravity ranges
  • Adjusted at 20°C
  • Includes protective plastic tube
  • Available for both low and medium surface tension
  • 250mm in length
  • Divided every 0.002g/mL
Specifications
Quantity1Pack
TypeM100 Density Hydrometer
Accuracy0.002g/mL
Increments0.002 g/mL
Certifications/ComplianceISO 387, ISO 649
Length (Metric)250 mm
IncludesPlastic protective tube
Ballast TypeSteel Shot
Baume Range1.2 to 1.3 g/mL
For Use With (Application)Adjusted for medium surface tension
Specific Gravity Range1.200 to 1.300 g/mL
Unit SizeEA
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link