Alcohol Burner
Alcohol Burner
Actual product may vary
Fisherbrand
Alcohol Burner
Enhance your experimentation using an Alcohol Burner with wick and cap when applying heat to laboratory substances.
Have Questions?
Catalog number FKS12812174
Price (SGD)
25.00
EA
Add to cart
Price (SGD)
25.00
EA
Add to cart
Enhance your experimentation using an Alcohol Burner with wick and cap when applying heat to laboratory substances.
  • With wick and cap
  • 100mL capacity
  • For burning methylated spirits during laboratory for experimentation
Specifications
Product TypeLamp
Capacity (Metric)100 mL
IncludesCap, Wick
No. per Pack1 Ea.
Weight (Metric)0.16356 kg
Unit SizeEA