Check Order Status
Test Tube Brushes with White Bristle Sponge End
Actual product may vary
Fisherbrand
Test Tube Brushes with White Bristle Sponge End
On galvanised steel wire handle. Pinnacle Brush™ Test Tube Brushes with White Bristle Sponge End is used for the cleaning of test tubes.
Have Questions?
Catalog Number
Color
FKS12811964
White
Catalog number
FKS12811964
Price (SGD)
8.00
EA
Add to cart
Color:
White
Price (SGD)
8.00
EA
Add to cart
Galvanised steel wire handle
Specifications
Product Type
Test Tube Brush
Handle Material
Steel Wire
Color
White
For Use With (Equipment)
Test tubes
Length (Metric)
265 mm
Quantity
1 Ea.
Size
55 x 40 mm