Burette Bristle Brushes
Burette Bristle Brushes
Actual product may vary
Fisher Catalogue
Burette Bristle Brushes
Excellent for cleaning out narrow burettes and other hard-to-reach instruments. Pinnacle Brush™ Burette Bristle Brushes are supplied in packs of 10.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric) BristleFor Use With (Equipment)
FKS1281194462 cm16 mm25 mL Burette
FKS1280194452 cm13 mm10 mL Burette
FKS1233832877 cm19 mm50 mL Burette
FKS1234832893 cm25 mm100 mL Burette
Catalog number FKS12811944
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric):
62 cm
Diameter (Metric) Bristle:
16 mm
For Use With (Equipment):
25 mL Burette
  • On galvanized steel wire handles
  • Available for a range of burette sizes and cleaning head diameters
Specifications
Product TypeBurette Brush
Bristle MaterialFiber
Handle MaterialMetal
Diameter (Metric)1.9 cm
Diameter (Metric) Bristle16 mm
For Use With (Equipment)25 mL Burette
Length (Metric)62 cm
Length (Metric) Brush10 cm
Length (Metric) Handle52 cm
Quantity10 Pk.
SizeBrush Circumference: 6 cm
Unit SizePK