Sterilin™ Polystyrene 30mL Universal Containers
Sterilin™ Polystyrene 30mL Universal Containers
Actual product may vary
Fisherbrand
Sterilin™ Polystyrene 30mL Universal Containers
Ensure excellent sample containment using Sterilin Polystyrene 30mL Universal Containers, featuring a traditional flow seal with 1-1/2 turn cap.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberIncludesLabelNo. per Case
FKS12799855Container, Boric AcidPrinted400/Cs.
FKS12501299Container OnlyNo Label400/Cs.
FKS12511299Container OnlyPrinted400/Cs.
FKS11768918Container, SpoonPrinted400/Cs.
Catalog number FKS12799855
Price (SGD)
648.00
PK
Add to cart
Includes:
Container, Boric Acid
Label:
Printed
No. per Case:
400/Cs.
Price (SGD)
648.00
PK
Add to cart
Ensure excellent sample containment using Thermo Scientific™ Sterilin Polystyrene 30mL Universal Containers, featuring a traditional flow seal with 1-1/2 turn cap.
NOT AVAILABLE IN AMERICA
  • Leak-tested in accordance with BS EN ISO 6717
  • 95kPa-compliance ensures leak-free performance and suitability for transport (except spoon variants)
  • Available pre-filled with boric acid (0.4g) 
  • Suitable for centrifugation at 3,800 x g
Ordering Alerts:
Not available in all geographic regions
Specifications
Product TypeUniversal Container
MaterialPolystyrene
ShapeCylindrical
Capacity (Metric)30 mL
Certifications/ComplianceLeak-tested in accordance with BS EN ISO 6717
Closure MaterialPolypropylene
DescriptionUniversal Container + boric acid
Diameter (Metric)31 mm
Height (Metric)94 mm
IncludesContainer, Boric Acid
LabelPrinted
No. per Case400/Cs.
No. per Pack50/Pk.
Unit SizePK