Silica Evaporating Basins
Product Image
Fisherbrand
Silica Evaporating Basins
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS1278738645 mL
FKS12707396100 mL
FKS1275738675 mL
Catalog number FKS12787386
Price (SGD)
247.00
EA
Add to cart
Capacity (Metric):
45 mL
Price (SGD)
247.00
EA
Add to cart
Specifications
Capacity (Metric)45 mL
MaterialSilica
ShapeRound
TypeEvaporating Basin
Height (Metric)27 mm
Diameter (Metric)75 mm
Unit SizeEA