Clifton™ Digital Thermostirrer Water Bath with Clear Copolyester Tank and Stainless Steel Shelf
Clifton™ Digital Thermostirrer Water Bath with Clear Copolyester Tank and Stainless Steel Shelf
Actual product may vary
Fisherbrand
Clifton™ Digital Thermostirrer Water Bath with Clear Copolyester Tank and Stainless Steel Shelf
Get maximum observation of samples, water circulation, and temperature stability with the stirred water bath. Designed for immersion of flasks, bottles, racks in a highly stable temperature environment, this Clifton™ Digital Thermostirrer Water Bath with Clear Copolyester Tank and Stainless Steel Shelf includes anti-bacterial technology and other unique features as demanded by many clinical and laboratory environments.
Have Questions?
Catalog NumberCapacity (Metric)
FKS1277890822 L
Catalog number FKS12778908
Price (SGD)
6,744.00
EA
Add to cart
Capacity (Metric):
22 L
Price (SGD)
6,744.00
EA
Add to cart
  • Stirred Clifton water bath designed for immersion of flasks, bottles, racks in a highly stable temperature environment
  • Digital PID temperature control
  • Sturdy clear co-polyester BPA free tank allows observation of samples
  • Thermostirrer controls and painted surfaces feature “anti-bacterial technology” 
  • When low liquid level is detected, a “FiLL” message automatically appears and heating is suspended
  • Every unit undergoes a factory 2 point calibration procedure 
Specifications
DescriptionDigital Water Bath
Capacity (Metric)22 L
Temperature Range5°C to 90°C
DisplayDigital LED
ModelNE4-22 D/CT
Temperature Uniformity±0.01°C
Warranty3 Years
Dimensions (D x W x H)325 x 530 x 345 mm
Dimensions (D x W x H) Interior290 x 385 x 200 mm
Width (Metric)530 mm
Width (Metric) Interior385 mm
Height (Metric)345 mm
Height (Metric) Interior200 mm
Depth (Metric) Interior290 mm
Weight (Metric)9 kg
Voltage230 V
Wattage1250 W