Clifton™ Stainless Steel Gable Lids
Clifton™ Stainless Steel Gable Lids
Actual product may vary
Fisherbrand
Clifton™ Stainless Steel Gable Lids
Obtain optimum results keeping samples in a water bath free from contamination and saving energy. Designed allowing for full use of whole width of a Clifton™ water bath, Clifton™ Stainless Steel Gable Lids help reduce energy consumption and running costs.
Have Questions?
Catalog NumberFor Use With (Equipment)
FKS1277888822 and 28 L Unstirred and Shaking Water Baths
Catalog number FKS12778888
Price (SGD)
996.00
PK
Add to cart
For Use With (Equipment):
22 and 28 L Unstirred and Shaking Water Baths
Price (SGD)
996.00
PK
Add to cart
  • Designed allowing full use of whole width of a Clifton™ water bath
  • Helps reduce evaporation
  • Asists in keeping samples in a water bath free from contamination and saves energy
  • Allows extended use of the bath, right into the edges of the tank
  • Lid design ensures all condensate returns back into the bath
  • Insulated handles for safe use
  • Reduces energy consumption and running costs for your Clifton water bath
  • Lowest height of the gable being 45mm above the bath
Specifications
For Use With (Equipment)22 and 28 L Unstirred and Shaking Water Baths