Clifton™ Stainless Steel Rectangular Electric Boiling Bath, 6 x 80mm Holes
Clifton™ Stainless Steel Rectangular Electric Boiling Bath, 6 x 80mm Holes
Actual product may vary
Fisherbrand
Clifton™ Stainless Steel Rectangular Electric Boiling Bath, 6 x 80mm Holes
Rectangular, stainless steel electrically heated boiling bath, with fixed temperature for boiling water, fitted with concentric ringed lids to vary the hole size.
Have Questions?
Catalog number FKS12778868
Price (SGD)
3,273.00
EA
Add to cart
Price (SGD)
3,273.00
EA
Add to cart
  • The steam produced provides an effective source of heat for samples:
    • that may be otherwise thermally damaged by a naked flame
    • used for applications to distil/concentrate liquids, melt agar and other solids
    • metallurgical analysis, protein analysis and bacteriological examinations
  • Features an integral constant level device to maintain constant water level during boiling and supplied with sets of rings to vary the diameter of each hole to accommodate different sizes of vessels of evaporating dishes and glassware, including round-bottom flasks simultaneously.
  • Fitted with a sheathed immersion element and a one shot thermal cut out for safety.
  • The bath provides constant boiling water for gentle heating of samples and in steam.
  • Accessory ER-1 Energy Regulator can be connected to boiling bath allowing control of the amount of boiling from a gentle simmer to a vigorous boil.
  • Always site the Boiling Bath on a heat proof surface.
  • Warranty: 2 year
Safety Features:
  • Always connect the constant level with laboratory tubing to a water supply, adjust the flow to gently top up the water level, whilst maintaining boiling.
  • The Constant water level has a visible water spout for checking water is topping up the Boiling bath in use and avoids the chances of boiling dry.
  • Manual reset cut out.
  • Always read the instruction book
Specifications
DescriptionStainless steel rectangular 6 hole boiling bath with concentric rings
TypeStainless Steel Rectangular 6 Hole Boiling Bath with Concentric Rings
IncludesConcentric rings
MaterialStainless Steel
Min. Opening Diameter (All Rings Removed)80 mm
No. of Positions6
Temperature (Metric) Maximum100°C
Warranty2 Years
Dimensions (L x W x H)321 x 219 x 90 mm
Width (Metric)321 mm
Diameter (Metric)30 mm
Shipping Weight (Metric)2.7 kg
Electrical Requirements230 V
Voltage230 V
Wattage1000 W