Check Order Status
Waste Sack Blue Plastic 457 x 737 x 965mm
Fisherbrand
Waste Sack Blue Plastic 457 x 737 x 965mm
Have Questions?
Catalog number
FKS12776328
Price (SGD)
-
10kg. Manufactured from 100% recycled material.
• Blue
• 457 x 737 x 965mm