Stuart™ TE-10D Circulator
Stuart™ TE-10D Circulator
Actual product may vary
Fisherbrand
Stuart™ TE-10D Circulator
Suitable for most routine laboratory applications
Have Questions?
Catalog number FKS12767929
Price (SGD)
7,878.00
EA
Add to cart
Price (SGD)
7,878.00
EA
Add to cart
Techne™ first introduced their classic clip-on thermoregulator back in 1948, and now offer this and three other similar units operating over different temperature ranges, all designed to be used with Techne™ unheated water baths or any other suitable laboratory vessel. They will all heat, circulate and safely control the temperature of the liquid in the bath within precisely controlled limits
  • Manufacturer part number FTE10DDC
  • Temperature range -40°C to 120°C (a separate cooling coil is required for sub-ambient temperatures)
  • Excellent temperature stability: ±0.01°C at 40°C
  • Four-digit setting with a bright LED digital temperature display
  • Suitable for most routine laboratory applications
  • User adjustable over-temperature cut-out
  • Low liquid level cut-out as standard
Specifications
Temperature Range-40°C to +120°C
TypeCirculator
Electrical Requirements120/240 V
Voltage120/240 V
Unit SizeEA