BioClean™ Excell BEXS Sterile Nitrile Cleanroom Gloves
BioClean™ Excell BEXS Sterile Nitrile Cleanroom Gloves
Actual product may vary
Fisher Catalogue
BioClean™ Excell BEXS Sterile Nitrile Cleanroom Gloves
Latex-free nitrile gloves are hand specific and resistant to a range of chemicals. Recommended for Class 10/ISO 4 critical environments.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
FKS127658356.5
FKS127458356
Catalog number FKS12765835
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Size:
6.5
  • 12 in. (30 cm) hand-specific
  • Sterile gamma irradiated nitrile
  • Nitrile is resistant to a wide-range of chemicals
  • Hypoallergenic
  • Anti-static properties
  • 1.5 freedom from pinholes
  • Powder free, Latex-free, beaded cuff, accelerator-free
  • Processed in a cleanroom environment to ensure compatibility with ISO Class 4 & EU GMP Grade A operating conditions
  • Sterility Assurance Level 10-6
Specifications
Certifications/ComplianceEU GMP Grade A, EN 420:2003, EN 455, EN 374-1, 2, 3:2003, ISO9001:2008, 89/686/EEC Article 11b, NEBB, ISO2859, PPE Directive 89/686/EEC
Certifications/Compliance (Europe)CE 0598, EN 420:2003 + A1:2009, EN 421:2010, EN ISO 374-1:2016, EN ISO 374-5:2016, EN 455 Part 2, Food
Cleanroom ClassClass 10 (ISO 4)
AQL1.5
ColorWhite
Exterior FinishFully Textured
Finger Thickness (English)6.6 mil
Finger Thickness (Metric)0.17 mm
HandSpecific
Latex FreeYes
MaterialNitrile
Palm Thickness (English)4.7 mil
Palm Thickness (Metric)0.12 mm
PowderedPowder Free
Size6.5
SterilitySterile
Thickness (English)6.6 mil
Thickness (Metric)0.17 mm
Manufacturing/QMS Audit StandardsISO 9001, PPE Regulation 2016 425 Module D
ModelBEXS65
SeriesBioClean™ BEXS
Length (English)12 in.
Length (Metric)30.5 cm
Unit SizePK