It is designed to cut PTFE, ETFE and polymers in general and PEEK capillary tubing in particular.
Have Questions?
Catalog number FKS12762680
Price (SGD)
96.00
EA
Add to cart
Price (SGD)
96.00
EA
Add to cart
Cuts PEEK, PTFE, ETFE, and another polymeric tubing
No distortion to OD or ID
Ideal for the most demanding LC applications
Safety lock secures the blade
Right angles and clean cuts are essential in LC to avoid leaks and dead volume.
This handy tool makes it easy to get burr-free, perpendicular ends on polymeric tubing without distorting the outside diameter and/or closing the inside diameter of the tubing.
The compact design and unique safety locking mechanism allow every chromatographer to keep one in their pocket.
Specifications
TypeClean-cut Tubing Cutter
DescriptionFor 1/16 in., 1/8 in., 2 mm, 3 mm, and 4 mm OD tubing