Precision Macro Cell Cuvettes with Lid
Precision Macro Cell Cuvettes with Lid
Actual product may vary
Precision Macro Cell Cuvettes with Lid
Precision Macro Cell Cuvettes with Lid
Fisherbrand
Precision Macro Cell Cuvettes with Lid
Designed to hold samples for spectroscopic experiments and optical analysis. Hellma™ Precision Macro Cell Cuvettes provide optically excellent transparency.
Have Questions?
Catalog NumberPath Length
FKS1276167510 mm
Catalog number FKS12761675
Price (SGD)
-
Path Length:
10 mm
  • With flat lid
  • Wavelength of 360 to 2500nm
  • Manufactured from optical glass
Specifications
TypeMacro Cell
Capacity (Metric)3.5 mL
MaterialOptical Glass
Quantity1 Pack
Path Length10 mm
Wavelength Range360 to 2500 nm
For Use With (Equipment)Spectro photo meter
For Use With (Application)Spectroscopy
ShapeRectangle
Unit SizeEA