Spectra/Por™ 1-4 Standard RC Dialysis Membrane in Flat Sheets
Spectra/Por™ 1-4 Standard RC Dialysis Membrane in Flat Sheets
Actual product may vary
Fisherbrand
Spectra/Por™ 1-4 Standard RC Dialysis Membrane in Flat Sheets
Clear, flexible and sturdy Standard RC (regenerated cellulose) in single layer square sheets that can be used with a variety of dialysis equipment or can be cut into specific dimensions for attaching to support frames for customized diagnostic assays, equilibrium studies, desalting, protein purification, etc.
Have Questions?
Catalog NumberMWCOWidth (English)Length (English)Membrane
FKS127575163.5 kDa4.2 in.4.2 in.Spectra/Por 3
Catalog number FKS12757516
Price (SGD)
1,824.00
PK
Add to cart
MWCO:
3.5 kDa
Width (English):
4.2 in.
Length (English):
4.2 in.
Membrane:
Spectra/Por 3
Price (SGD)
1,824.00
PK
Add to cart
  • Flat-Sheets deal for use with dialysis equipment or cutting to size
  • Available in 3.5 kD, 6-8 kD and 12-14 kD MWCO's
  • Good chemical compatibility with dilute strong acids & bases, conc. weak acids and bases, many alcohols & some organics.
  • Standard RC can tolerate pH 2 - 12 and temp 4-121°C.
  • 25 sheets/pkg
Do not tie knots in tubing; use closures
Specifications
Product TypeSpectra/Por 3 Dialysis Membrane Flat Sheets
MWCO3.5 kDa
MembraneSpectra/Por 3
Length (English)4.2 in.
Length (Metric)108 mm
Width (English)4.2 in.
Width (Metric)108 mm
Dimensions (L x W)4.2 x 4.2 in. (108 x 108 mm)
ColorClear
Temperature Range (English)39°F to 250°F
Temperature Range (Metric)4°C to 121°C
Quantity25 Sheets
For Use With (Application)Ideal for molecular separation, protein binding studies, electrolyte removal, desalting
Unit SizePK