BioClean™ Extra BLAS Sterile Latex Cleanroom Gloves
BioClean™ Extra BLAS Sterile Latex Cleanroom Gloves
Actual product may vary
Fisherbrand
BioClean™ Extra BLAS Sterile Latex Cleanroom Gloves
Powder free, sterile gloves are flexible and extra long to provide elbow length protection. Recommended for Class 10/ISO 4 Cleanrooms.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
FKS12755825Large
FIS19-668-076Small
FKS12735825Small
FIS19-668-077Medium
FKS12745825Medium
FIS19-668-078Large
Catalog number FKS12755825
Price (SGD)
-
Size:
Large
  • Low levels of latex proteins
  • Extended length 16 in. (40 cm)
  • Ambidextrous, beaded cuff
  • Powder free
  • Gamma irradiated
  • Fully textured for improved grip
  • Flexibile and comfortable
  • Easy double-donning
Specifications
Certifications/ComplianceASTM D3578
Certifications/Compliance (Europe)EN 420:2003 + A1:2009 Category III, EN 455 Part 2, EN374-1:20016, EN374-5:2016 and EN421:2010, ISO 11137-1:2006, (EC)1935/2004 and (EC)2023/2006
Cleanroom ClassClass 10 (ISO 4)
AQL1.5
ColorNatural
Exterior FinishFully Textured
Finger Thickness (English)8.2 mil
Finger Thickness (Metric)0.21 mm
HandAmbidextrous
MaterialLatex
Palm Thickness (English)6.6 mil
Palm Thickness (Metric)0.17 mm
PowderedPowder Free
SizeLarge
SterilitySterile
Thickness (English)8.2 mil
Thickness (Metric)0.21 mm
Manufacturing/QMS Audit StandardsISO 9001, PPE Regulation 2016 425 Module D
ModelBLAS-L
SeriesBioClean™ Extra BLAS
Length (English)16 in.
Length (Metric)40 cm
Unit SizePK