Nelldorn Glass Dryer
Nelldorn Glass Dryer
Actual product may vary
Nelldorn Glass Dryer
Nelldorn Glass Dryer
Nelldorn Glass Dryer
Fisherbrand
Nelldorn Glass Dryer
An economical and efficient method of drying and storing most containers, glassware and plasticware.
Have Questions?
Catalog NumberHeight (Metric)Width (Metric)
FKS12753015430 mm490 mm
Catalog number FKS12753015
Price (SGD)
12,308.00
EA
Add to cart
Height (Metric):
430 mm
Width (Metric):
490 mm
Price (SGD)
12,308.00
EA
Add to cart
  • Unit can be used for most types of dispensing containers including plastics
  • Equipment can be left on the rack until drying is complete
  • Capable of holding up to 60 items with 15-20 minutes drying times
  • Wall mountable or free-standing
  • Thermostatically controlled
  • Economical, quick and efficient
  • Comes with a variable 120-minute timer
  • A large volume of air is drawn over the controlled heating elements and forced through each drying tube
  • Rapid flow of air dries glass or plastic quickly and without any damage to the apparatus
  • Very low power consumption
  • Stepped design allows the removal of any item without disturbing the remainder of the load and facilitates the run-off of water to the front of the unit where a small drain pipe prevents it collecting at the base
  • Air intake is recessed at the back of the unit to allow wall or bench mounting
  • Fitted with 60 rigid stain resistant nylon drying tubes with an adjustable timer which can be preset to switch off at your desired time
Specifications
Product TypeDrying Rack
For Use With (Application)Can Hold 30 x 75 mm Tubes, 13 x 100 mm Tubes, 9 x 40 mm Tubes, and 8 x 152 mm Tubes
IncludesVariable 120 minute timer
Height (Metric)430 mm
Width (Metric)490 mm
Voltage220/240 V
Frequency50 Hz
Unit SizeEA