Stainless Steel Blunt End Spring Forceps
Product Image
Fisherbrand
Stainless Steel Blunt End Spring Forceps
Use these forceps for a variety of life science applications. S Murray™ Stainless Steel Blunt End Spring Forceps are fully guaranteed in normal use and subject to stringent quality inspection procedures.
Have Questions?
Catalog number FKS12750916
Price (SGD)
18.00
PK
Add to cart
Price (SGD)
18.00
PK
Add to cart
Use these forceps for a variety of life science applications. S Murray™ Stainless Steel Blunt End Spring Forceps are fully guaranteed in normal use and subject to stringent quality inspection procedures.
  • Standard instruments are finished to a bright mirror polish
  • Stainless Steel
  • Length: 200mm
Specifications
Product TypeSpring Forceps
MaterialStainless Steel
For Use With (Application)For dissecting
Tip StyleBlunt
Length (Metric)200 mm
Unit SizePK