Clifton™ Stainless Steel Raised Shelf
Clifton™ Stainless Steel Raised Shelf
Actual product may vary
Fisherbrand
Clifton™ Stainless Steel Raised Shelf
Obtain a good even heat distibution with perforted shelves. Nickel Electro™ Clifton™ Stainless Steel Raised Shelf allows two immersion depths by inverting the shelf.
Have Questions?
Catalog NumberSizeFor Use With (Equipment)
FKS1274889830 x 30 x 20 cm14, 22, 28 And 38 Liter Baths
Catalog number FKS12748898
Price (SGD)
516.00
EA
Add to cart
Size:
30 x 30 x 20 cm
For Use With (Equipment):
14, 22, 28 And 38 Liter Baths
Price (SGD)
516.00
EA
Add to cart
  • Simple solution for placing shallow vessels in a stirred bath
  • Manufactured from stainless steel
  • The raised shelf is perforated to allow free circulation of water
  • Can be easily removed and cleaned
  • Shelf can be inverted to provide two different immersion depths
Specifications
For Use With (Equipment)14, 22, 28 And 38 Liter Baths
TypeShelf Raised
MaterialStainless Steel
Size30 x 30 x 20 cm
Dimensions (L x W x H)30 x 30 x 20 cm
Unit SizeEA