M-Zone™ Bottle Brush
M-Zone™ Bottle Brush
Actual product may vary
Fisherbrand
M-Zone™ Bottle Brush
Color-coded bottle brushes with rounded, stiff polyester bristles for versatile and aggressive cleaning of bottles and various equipment in production and in the lab.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorDiameter (Metric) Bristle
FKS12735755White7.6 cm
FKS12725755White6.4 cm
Catalog number FKS12735755
Price (SGD)
270.00
EA
Add to cart
Color:
White
Diameter (Metric) Bristle:
7.6 cm
Price (SGD)
270.00
EA
Add to cart
A range of color-coded bottle brushes with rounded, stiff polyester bristles for versatile and aggressive cleaning of bottles and various equipment in production and in the lab.
The Micronova M-Zone™ Bottle Brushes are multipurpose brushes with rounded, stiff polyester bristles and either 12 or 16-inch plastic easy grip handles. Designed to clean bottles and other lab and production equipment.
  • Multipurpose, color-coded plastic bottle brushes
  • Durable, stiff polyester bristles
  • Easy grip plastic handle
  • For versatile and aggressive cleaning of bottles and equipment
  • Made from FDA approved, BPA free base resins
  • Available in Blue, Green, Red, White and Yellow
Specifications
Product TypeBottle Brush
Bristle MaterialPolyester
Bristle TypeSoft
Color-codedNo
Handle MaterialPolypropylene
Handle TypeCurved
Tip TypeCurved
SterilityNon-sterile
AppearanceRounded, white, Bottle Brush Head with soft bristles, attached to a fixed length, white, plastic handle with rounded end.
AutoclavableNot Autoclavable
Care InstructionsKeep at ambient temperature
Certifications/ComplianceCertificate of Conformance; ISO 9001 Certified Facility; FDA Approved
ColorWhite
Diameter (English) Bristle3 in.
Diameter (Metric) Bristle7.6 cm
Dimensions (L x W x H)16 x 3 x 3 in. (40.6 x 7.6 x 7.6 cm)
DisposableReusable
FilteredNon-filtered
FlammableNo
For Use With (Application)Color-Coded Bottle Brushes for Equipment Cleaning
Height (English)3 in.
Height (Metric)7.6 cm
Length (English)16 in.
Length (English) Bristle3 in.
Length (English) Brush7 in.
Length (English) Handle9 in.
Length (Metric)40.6 cm
Length (Metric) Bristle7.6 cm
Length (Metric) Brush17.7 cm
Length (Metric) Handle22.9 cm
MaterialPolypropylene, Polyester
PackagingEach brush single bagged
Quantity12/Cs.
Resistant ToStandard disinfectants and cleaning solutions
Size16 x 10 x 6 in.
Width (English) Bristle3 in.
Width (Metric) Bristle7.6 cm
Unit SizeEA
Contents & Storage
Keep at ambient temperature