Stainless Steel Forceps
Stainless Steel Forceps
Actual product may vary
Fisherbrand
Stainless Steel Forceps
Use these forceps for a wide range of applications. S Murray™ Stainless Steel Forceps are made from only high grade stainless steel in accordance to the specifications of ISO 7153-1.
Have Questions?
Catalog NumberLength (Metric)Tip StyleFor Use With (Application)
FKS12730046125 mmBlocked end, Plain serrated jaws, StraightDissection applications
Catalog number FKS12730046
Price (SGD)
11.00
EA
Add to cart
Length (Metric):
125 mm
Tip Style:
Blocked end, Plain serrated jaws, Straight
For Use With (Application):
Dissection applications
Price (SGD)
11.00
EA
Add to cart
  • Stainless Steel forceps
Specifications
Product TypeForceps
MaterialStainless Steel
For Use With (Application)Dissection applications
QuantityEach
Tip StyleBlocked end, Plain serrated jaws, Straight
Length (Metric)125 mm
Unit SizeEA