Buffer Solutions: Standard pH
Buffer Solutions: Standard pH
Actual product may vary
Fisherbrand
Buffer Solutions: Standard pH
Storage and refill solutions for pH electrodes
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorpH RangePackaging
FKS12723161Clear, Green730 x 20 mL Sachets
FKS12663516Clear, Red430 x 20 mL Sachets
Catalog number FKS12723161
Price (SGD)
385.00
PK
Add to cart
Color:
Clear, Green
pH Range:
7
Packaging:
30 x 20 mL Sachets
Price (SGD)
385.00
PK
Add to cart
  • Completely miscible
  • Comes with Quality inspection certificate
  • NIST traceable
Specifications
Boiling Point∼100°C
ColorClear, Green
TypeBuffer Solution
Filling Solution TypepH Buffer
IncludesQuality inspection certificate
Packaging30 x 20 mL Sachets
pH Range7
Water SolubleCompletely Miscible
Unit SizePK