Stainless Steel Dissection Dish
Stainless Steel Dissection Dish
Actual product may vary
Fisherbrand
Stainless Steel Dissection Dish
Use a dissecting dish when working with and transporting small specimens. Nickel Electro™ Stainless Steel Dissection Dish is made of durable stainless steel and comes in various sizes.
Have Questions?
Catalog NumberDimensions (L x W x H)
FKS12720036300 x 450 x 50 mm
Catalog number FKS12720036
Price (SGD)
329.00
EA
Add to cart
Dimensions (L x W x H):
300 x 450 x 50 mm
Price (SGD)
329.00
EA
Add to cart
  • Stainless steel
  • Shallow form
  • One piece pressing
  • Useful when working with and transporting small specimens
Specifications
Bottom ShapeFlat
MaterialStainless Steel
ShapeRectangular
TypeDissection Dish
Dimensions (L x W x H)300 x 450 x 50 mm
Length (Metric)300 mm
Width (Metric)450 mm
Height (Metric)50 mm
Unit SizeEA