Check Order Status
Shandon™ Stainless-Steel Scalpel Blade Handle, Holds No. 20-25 Blades
Actual product may vary
Fisherbrand
Shandon™ Stainless-Steel Scalpel Blade Handle, Holds No. 20-25 Blades
Securely hold blades with the Epredia™ Shandon™ Stainless-Steel Scalpel Blade Handle, constructed of #4 stainless steel.
Have Questions?
Catalog Number
Size
FKS12697746
# 4
Catalog number
FKS12697746
Price (SGD)
234.00
EA
Add to cart
Size:
# 4
Price (SGD)
234.00
EA
Add to cart
Properly hardened, tempered and passivated
Electronically polished and cleaned for a high-quality finish
Quality patterns, dimensions and finish held to exacting standards, producing the finest instruments available
Defect-free in material and workmanship
Specifications
Blade Material
Stainless Steel
Size
# 4
Length (English)
8.5 in.
Length (Metric)
21.6 cm
For Use With (Equipment)
Holds # 20 to # 25 Blade