Check Order Status
Coplin Staining Jar, Holds ten slides back-to-back
Actual product may vary
Fisherbrand
Coplin Staining Jar, Holds ten slides back-to-back
Hold 10 slides back-to-back with the Epredia™ Coplin Staining Jar.
Have Questions?
Catalog Number
Slide Count
FKS12697696
10
Catalog number
FKS12697696
Price (SGD)
258.00
EA
Add to cart
Slide Count:
10
Price (SGD)
258.00
EA
Add to cart
Features:
1.6in. (40mm) diameter circular, tapered top
Heavy, clear glass jar with internal grooves
Holds ten slides (76 x 26 mm) back-to-back
Includes:
Plastic screw-on cap.
Specifications
Closure Included
Closure Included
Bottom Shape
Flat
Material
Glass
Type
Coplin Staining Jar
Slide Count
10
Diameter (English)
16 in.
Diameter (Metric)
40 mm