Nunc™ Sterile Flask Replacement Filter Caps
Nunc™ Sterile Flask Replacement Filter Caps
Actual product may vary
Fisherbrand
Nunc™ Sterile Flask Replacement Filter Caps
Allow constant airflow and minimize chance of contamination by using individually packed Thermo Scientific™ Nunc™ Sterile Flask Replacement Filter Caps.
Have Questions?
Catalog NumberFor Use With (Equipment)
FKS12696856Nunc EasYFlasks 75 cm²
Catalog number FKS12696856
Price (SGD)
354.00
PK
Add to cart
For Use With (Equipment):
Nunc EasYFlasks 75 cm²
Price (SGD)
354.00
PK
Add to cart
Allow constant airflow and minimize chance of contamination with Thermo Scientific™ Nunc™ Sterile Flask Replacement Filter Caps.
Nunc sterile filter caps are made with HDPE and Thermo Scientific™ GORE™ dPTFE membrane.
  • Filter membrane provides effective barrier against microorganisms
  • Chemically robust membrane meets USP class VI standards and is produced without animal-derived substances
  • Well-defined, hydrophobic filter membrane offers continuous venting with consistent gas exchange and no lot-to-lot variation
  • Sterile
Warranty
: 90 days
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Closure ColorWhite
MaterialHDPE
For Use With (Equipment)Nunc EasYFlasks 75 cm²
No. per Case100
No. per Pack1
SterilitySterile
ClassClass VI
Closure TypeFilter
Culture Area75 cm2
Unit SizePK