Stainless Steel Laboratory Bottle, with DIN 168-1 Thread, GL 45
Stainless Steel Laboratory Bottle, with DIN 168-1 Thread, GL 45
Actual product may vary
Fisherbrand
Stainless Steel Laboratory Bottle, with DIN 168-1 Thread, GL 45
Corrosion and damage resistant stainless steel laboratory bottle.
Have Questions?
Catalog NumberCapacity (Metric)
FKS126961671500 mL
Catalog number FKS12696167
Price (SGD)
2,517.00
EA
Add to cart
Capacity (Metric):
1500 mL
Price (SGD)
2,517.00
EA
Add to cart
The unbreakable steel bottle is ideal for storage applications where glass is not applicable, due to the risk of breakage or chemical incompatibility. Manufactured from corrosion resistant AISI Type 316L (1.4404) Stainless steel, and hygienically constructed with all welded seams. The bottle has a smooth inner surface finish that corresponds to IIIc (DIN 17441) with rounded inner edges for ease of cleaning. Polished and brushed durable exterior finish. The bottle has a GL 45 thread with a built-in pouring lip. Supplied without a cap, Stainless steel cap is available. It is fully compatible with all the GL 45 caps.
  • Manufactured from corrosion resistant AISI Type 316L (1.4404) Stainless steel, and hygienically constructed with all welded seams
  • Polished and brushed durable exterior finish
  • Due to the smooth inner surface that corresponds to IIIc (DIN 17441) with rounded inner edges the bottle can be easily cleaned
  • GL 45 thread with a built-in pouring lip
  • Fully compatible with all the GL 45 caps
  • Standardised GL-thread and matching cap system for particularly tight sealing and simple, clean pouring out
Specifications
Capacity (Metric)1500 mL
For Use With (Application)Storage, transport and safe handling of hazardous, dangerous and valuable materials; Storage and transport of light-sensitive materials
Unit SizeEA