Traceable™ LCD Timer with Nano Counter
Traceable™ LCD Timer with Nano Counter
Actual product may vary
Fisherbrand
Traceable™ LCD Timer with Nano Counter
Displays countdown alarm time/stopwatch (count-up) time and time of day. Fisherbrand™ LCD Timer with Nano Counter includes memory feature for repetitive use, large, easy-to-read digits, clip, stand and magnet on back.
Have Questions?
Catalog number FKS12695296
Price (SGD)
50.00
EA
Add to cart
Price (SGD)
50.00
EA
Add to cart
  • Two timing channels, each with 23 hours, 59 minutes, 59 seconds capacity
  • Clock works on AM/PM or 24-hour format
  • 60-second loud alarm
  • Memory feature for repetitive use
  • Large, easy-to-read digits
  • Clip, stand and magnet on back
Specifications
Product TypeClock/Timer
Time Range23 hr. 59 min. 59 sec.
Alarms60 sec. Loud Alarm
DisplayDigital
Dimensions (L x W x H)2.5 x 2 x 0.88 in. (6.35 x 5.08 x 2.23 cm)
Length (English)2.5 in.
Length (Metric)6.35 cm
Width (English)2 in.
Width (Metric)5.08 cm
Height (English)0.88 in.
Height (Metric)2.23 cm
Weight (English)0.2 lb.
Weight (Metric)0.09 kg
IncludesTwo Year battery, clip, stand, magnet on back
Unit SizeEA