Check Order Status
Gas Delivery Tube
Actual product may vary
Fisherbrand
Gas Delivery Tube
Polypropylene construction. Azlon™ Gas Delivery Tube are ideal substitutes for fragile glass alternatives.
Have Questions?
Change view
Catalog Number
Overall Length (Metric)
Product Type
FKS12693012
475 mm
Double Bend Gas Delivery Tube
FKS12673012
290 mm
Bung Top Gas Delivery Tube
2 Options
Catalog number
FKS12693012
Price (SGD)
127.00
PK
Add to cart
Overall Length (Metric):
475 mm
Product Type:
Double Bend Gas Delivery Tube
Price (SGD)
127.00
PK
Add to cart
Pre-bent rigid polypropylene delivery tubes
Unbreakable in normal use
Suitable for many types of gas collection experiments in schools and universities
Specifications
Description
Double bend
Diameter (Metric) Inner
4 mm
Material
Polypropylene
Diameter (Metric) Outer
6 mm
Overall Length (Metric)
475 mm
For Use With (Application)
Many types of gas collection experiments in schools and universities
Max. Operating Temperature (Metric)
135°C
Product Type
Double Bend Gas Delivery Tube