S 50 R Ultrasonic Bath
S 50 R Ultrasonic Bath
Actual product may vary
Fisherbrand
S 50 R Ultrasonic Bath
Fisherbrand™ S 50 R Ultrasonic Water Bath features microprocessor-controlled ultrasonic modes and special predefined programs to assist every standard laboratory and cleaning application.
Have Questions?
Catalog number FKS12689345
Price (SGD)
3,958.00
PK
Add to cart
Price (SGD)
3,958.00
PK
Add to cart
  • Plastic carrying handles
  • Detachable mains cable
  • User-friendly clearly arranged operating panel
  • Connection rack for laboratory holder, noise-reduction operation
  • Ultrasonic tank made of cavitation-resistant stainless steel
  • Special efficient and noise-reduced Degas mode (gas liftout effect)
  • Turning knob for setting continued and short-period operation from 1 to 30 min
  • LED display for both pre-set and actual temperature, and for remaining operating time
Specifications
Voltage230 VAC
Capacity (Metric)5.3 L
Dimensions (D x W x H)335 x 304 mm (H x Dia.)
Electrical Requirements230VAC, 150w
Height (Metric) Exterior335 mm
Height (Metric) Interior335 mm
ModelElmasonic S 50 R
Wattage150 w
Unit SizePK