Eutech™ ECOECTESTHIGH Conductivity Meter
Eutech™ ECOECTESTHIGH Conductivity Meter
Actual product may vary
Fisherbrand
Eutech™ ECOECTESTHIGH Conductivity Meter
The Eutech EcoTestr EC series comes with rugged, sturdy stainless steel pin sensors, giving you quick, reliable conductivity measurements.
Have Questions?
Catalog NumberMeasurement Modes
FKS12685915Conductivity
Catalog number FKS12685915
Price (SGD)
256.00
EA
Add to cart
Measurement Modes:
Conductivity
Price (SGD)
256.00
EA
Add to cart
The Eutech EcoTestr EC series comes with rugged, sturdy stainless steel pin sensors, giving you quick, reliable conductivity measurements. Ideal for use in a wide range of conductivity applications, such as hydroponics gardening, fish-farming, pools, electroplating and other water/ wastewater applications. Manual calibration option for better accuracy with near-to-sample calibrations. Click-lock battery compartment requires no tools for battery replacements. Hold function freezes reading temporarily for ease of recording. Sensor protective cap that doubles up as a container for samples or calibration solution.
Specifications
  • Range: .. to 19.90mS/cm
  • Resolution: 0.1mS/cm
  • Accuracy: ±1 % full scale (±2 % above 10mS/cm)
  • Calibration points: 1 (digital, push-button)
  • ATC: Yes
  • Non-Volatilememory: Yes
  • Hold Function: Yes
  • Auto-Off: 8.5mins after last key pressed
  • LCD Display: 3 ½ digit with annunciator
  • Power: 4 X 1.5 V 'A76'micro alkaline batteries
  • Ingress Rating: IP67 Waterproof
  • Dimensions; Weight: Boxed: 24.5 cm x 13.5 cm x 4.5 cm; 137 g
  • Specifications
    Accuracy (Conductivity)±1% Full Scale (±2% above 10 mS/cm)
    DescriptionWaterproof EcoTestr EC High with ATC; 1 point Calibration
    Measurement ModesConductivity
    Range (Conductivity)0 mS/cm to 19.9 mS/cm
    Display TypeLCD
    IP RatingIP67
    Unit SizeEA