Polypropylene Low Form Beakers with Blue Printed Graduations
Polypropylene Low Form Beakers with Blue Printed Graduations
Actual product may vary
Fisher Catalogue
Polypropylene Low Form Beakers with Blue Printed Graduations
Manufactured from highly translucent polypropylene for durability, clarity and good chemical resistance. Autoclavable Kartell™ Polypropylene Low Form Beakers with Printed Blue Graduations provide an economical and safe solution for stirring, mixing and dispensing liquids in the laboratory.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS12685686100 mL
FKS12625696250 mL
Catalog number FKS12685686
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
100 mL
Manufactured from highly translucent polypropylene for durability, clarity and good chemical resistance. Autoclavable Kartell™ Polypropylene Low Form Beakers with Printed Blue Graduations provide an economical and safe solution for stirring, mixing and dispensing liquids in the laboratory.
  • Permanent printed blue graduations provide clarity and reproducibility
  • Autoclavable at 121°C for 10 minutes
  • Withstands 100°C of continuous work
  • Class B, Low form
  • Complies with ISO 7056-1981(E) and BS 5404 Part 1
  • Suitable for foodstuff
Specifications
ColorTranslucent
Diameter (Metric) Outer51 mm
Capacity (Metric)100 mL
MaterialPolypropylene
For Use With (Application)Suitable for Foodstuff
Height (Metric)72 mm
Increments5 mL
Quantity12Pk.
Temperature (Metric) Max. Operating100°C for continuous work
TypeBeaker
AutoclavableAutoclavable
GraduatedYes
Certifications/ComplianceISO 7056-1981(E), BS 5404 Part 1
StyleLow Form
Unit SizePK