Armadillo PCR Plate, 384-well, clear, clear wells
Armadillo PCR Plate, 384-well, clear, clear wells
Actual product may vary
Fisherbrand
Armadillo PCR Plate, 384-well, clear, clear wells
Combine the rigidity of a polycarbonate frame with thin walled polypropylene wells to provide superior thermal cycling performance under all conditions without warping.
Have Questions?
Catalog NumberQuantity
FKS1268098550 plates
Catalog number FKS12680985
Price (SGD)
1,376.00
PK
Add to cart
Quantity:
50 plates
Price (SGD)
1,376.00
PK
Add to cart
Thermo Scientific™ Armadillo PCR plates combine the rigidity of a polycarbonate frame with thin walled polypropylene wells to provide superior thermal cycling performance under all conditions without warping.
Thermo Scientific Armadillo PCR plates combine the rigidity of a polycarbonate frame with thin-walled polypropylene wells to provide superior thermal cycling performance under all conditions without warping.
They are available in 96- and 384-well formats, in multiple colors, and can be ordered with a standard 128 barcode or custom barcoding. All formats are available with clear or white wells; choose white for superior qPCR results. The specially-designed warp resistant frame and multiple format options make Armadillo PCR plates the ultimate choice for high-throughput PCR and automated handling.
  • Polycarbonate frame for warp resistant thermal cycling
  • Enhanced mechanical stability for robotic handling
  • Thin walled polypropylene wells for optimal heat transfer
  • Well shape optimized for maximum sample recovery
  • Flat alpha-numeric lettering and raised rim well design for improved heat sealing
  • Optically clear deck allows for easy visibility of wells
  • Multiple frame color options, all available in both clear (for PCR) and white colored wells (for qPCR)
  • Maximum well volume 40μL
  • Maximum working volume 25μL
This product is sold under license from Eppendorf AG.
Specifications
BarcodeNo Barcode
Certifications/ComplianceCertified RNase/DNase-free
Packaging50 Plate
Product LineArmadillo
ColorClear
Product TypePCR Microplate
For Use With (Equipment)Real-Time PCR Systems or Thermal Cyclers
Skirt StyleSkirted
MaterialPolycarbonate, Polypropylene
Volume (Metric) Well40 μL
No. of Wells384
Volume (Metric) Working25 μL
Quantity50 plates
Reaction SpeedStandard
Well ColorClear
Unit SizePK