Sterilin™ Polypropylene 30mL Universal Containers
Sterilin™ Polypropylene 30mL Universal Containers
Actual product may vary
Fisherbrand
Sterilin™ Polypropylene 30mL Universal Containers
Secure excellent temperature and chemical resistance with the 95kPa compliant and CE marked 30mL leak-resistant container with Quickstart 1/2 turn cap.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLabelIncludes
FKS12673546PlainContainer Only
FKS12663546PrintedContainer Only
128CPPlainContainer Only
Catalog number FKS12673546
Price (SGD)
534.00
PK
Add to cart
Label:
Plain
Includes:
Container Only
Price (SGD)
534.00
PK
Add to cart
Secure excellent temperature and chemical resistance with these 95kPa compliant 30mL leak-resistant Thermo Scientific™ Sterilin™ Polypropylene 30mL Universal Containers.
NOT AVAILABLE IN AMERICA
Features:
  • Leak-tested in accordance with BS EN ISO 6717
  • Recommended for centrifugation at 3,800 x g
  • Multi-seal cap 'QuickStart' for improved handling and unrivaled leak-free performance
  • Lot number printed on each container to aid full traceability
  • Labeled and available either undosed, dosed with boric acid preservative (0.4g)
Ordering Alerts:
Not available in all geographic regions
Specifications
Product TypeContainer
MaterialPolypropylene
ShapeCylindrical
SterilityNon-sterile
Capacity (Metric)30 mL
Certifications/ComplianceLeak-tested in accordance with BS EN ISO 6717. 
Closure TypeMulti-Seal
DescriptionUniversal Container
Diameter (Metric)31 mm
Height (Metric)94 mm
IncludesContainer Only
LabelPlain
No. per Case400/Cs.
Unit SizePK