Eutech™ ECFC7252101B pH electrode
Product Image
Fisherbrand
Eutech™ ECFC7252101B pH electrode
Eutech™ offers a wide range of pH meters and electrodes that measure pH values in various educational, laboratory and industrial applications.
Have Questions?
Catalog NumberConnector Types
FKS12673125BNC
Catalog number FKS12673125
Price (SGD)
340.00
PK
Add to cart
Connector Types:
BNC
Price (SGD)
340.00
PK
Add to cart
The measure of a liquid acidity and alkalinity, pH is one of the most common parameter measured in a wide variety of industries ranging from water and wastewater treatment, chemical production, agriculture research and production, environmental monitoring, chemical and life sciences research, biochemical and pharmaceutical research, electronics production to food processing and other industrial applications. Eutech™ offers a wide range of pH meters and electrodes that measure pH values in various educational, laboratory and industrial applications. This open pore glass-body pH combination electrode is used for general pH measurements. Comes with 10 ml refilling electrolyte.
Features:
  • Parameter: pH
  • Range:0 to 13 pH
  • Temperature Range: 0 to 80 °C
  • Liquid Junction Type: Porous HDPE pin
  • Internal Reference Type: Ag/AgCl
  • Sealed/Refillable: Sealed
  • Reference Junction: Single
  • Dimensions (Shaft): 90 x 12mm
  • Cable Length: 1m
  • Connector: BNC
  • Used With: All pHmeters with BNC input connecto
Specifications
Connector TypesBNC
JunctionSingle
MaterialGlass Body
pH Range0 to 13
Temperature (Metric) Operating0°C to 80°C
TypepH Electrode
Unit SizePK