1x1 image pixel for data collection
EasyDip™ Slide Staining System Kit
EasyDip™ Slide Staining System Kit
Actual product may vary
EasyDip™ Slide Staining System Kit
EasyDip™ Slide Staining System Kit
EasyDip™ Slide Staining System Kit
Fisherbrand
EasyDip™ Slide Staining System Kit
User-friendly approach for staining microscope slides. These EasyDip™ Slide Staining Jars are a part of the EasyDip™ Slide Staining System. (See also the EasyDip Slide Staining Rack.)
Have Questions?
Catalog NumberSlide Count
FKS1266867512 standard microscope slides
Catalog number FKS12668675
Price (SGD)
328.00
PK
Add to cart
Slide Count:
12 standard microscope slides
Price (SGD)
328.00
PK
Add to cart
  • Slide Staining System Kit
  • This kit includes 5 jars (one of each color) and 1 rack (M905-12DGY)
  • Resistant acetal acid
  • Lid to minimize spills or evaporation
  • Handle attached to rack for easy insertion and removal
  • Wide stable base
  • Easy to clean
  • Not corroded by metals
  • Dimensions: 42.5 x 10.2 x 3.8 cm
Specifications
AutoclavableNot Autoclavable
Bottom ShapeFlat
Capacity (Metric)80 mL
ColorAssorted
MaterialAcetal
TypeSlide Staining Jar
For Use With (Application)Special stains, frozen sections and special processes
QuantitySet, 5 Pk.
Slide Count12 standard microscope slides
Length (English)3 in.
Length (Metric)76 mm
Width (English)2.5 in.
Width (Metric)64 mm
Height (Metric)92 mm
Height (English)3.625 in.
Unit SizePK
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link