Portex™ Fine Bore LDPE Tubing
Portex™ Fine Bore LDPE Tubing
Actual product may vary
Fisherbrand
Portex™ Fine Bore LDPE Tubing
Offering softness, a light weight and excellent chemical resistance in the laboratory. Smiths Medical™ Portex™ Fine Bore LDPE Tubing has a smooth bore for ease of cleaning.
Have Questions?
Catalog NumberDiameter (Metric) Inner
FKS126654970.58 mm
Catalog number FKS12665497
Price (SGD)
-
Diameter (Metric) Inner:
0.58 mm
  • Lightweight and flexible
  • Translucent in color
  • Surgical non-toxic quality, minimises trauma when used in vivo
  • Suitable for sheathing glass fibre in telecommunications
Specifications
AutoclavableNot Autoclavable
Diameter (Metric) Inner0.58 mm
MaterialPolyethylene
Diameter (Metric) Outer0.96 mm
Overall Length (Metric)30 m
Elongation450%
For Use With (Application)For sheathing delicate glass fibers in telecommunications
Min. Operating Temperature (Metric)-20°C
Packaging Quantity1 Pack
SterilitySterile
ColorTranslucent
FlexibilityFlexible
Maximum Pressure93 psi
Max. Operating Temperature (Metric)80°C
Unit SizeEA