Check Order Status
Polystyrene Petri Dishes
Actual product may vary
Fisherbrand
Polystyrene Petri Dishes
Clear. Fisherbrand™ Polystyrene Petri Dishes are perfect for microbial or cell culture applications.
Have Questions?
Change view
Catalog Number
Vent
FKS12664785
3
FKS12654785
1
2 Options
Catalog number
FKS12664785
Price (SGD)
243.00
PK
Add to cart
Vent:
3
Price (SGD)
243.00
PK
Add to cart
Specifications
Material
Polystyrene
Sterility
Non-sterile
Type
Petri Dish
Quantity
600 Case
Vent
3
Height (Metric)
16.2 mm
Diameter (Metric)
90 mm